U.S. Stainless Steel Test Sieve Covers
U.S. Stainless Steel Test Sieve Covers
Actual product may vary
U.S. Stainless Steel Test Sieve Covers
Cover, Sieve; Fisherbrand; For U.S. standard test sieve; Stainless Steel; For 12 in. dia. (30.5cm) sieves
Have Questions?
Catalog NumberDiameter (English) Outer
FIS04-888B12 in.
Catalog number FIS04-888B
Price (SGD)
225.00
EA
Add to cart
Diameter (English) Outer:
12 in.
Price (SGD)
225.00
EA
Add to cart
Cover, Sieve; Fisherbrand; For U.S. standard test sieve; Stainless Steel; For 12 in. dia. (30.5cm) sieves
Fit all 8 and 12 in. diameter (20.3 and 30.5cm) sieves.
Specifications
DescriptionTop Cover with Ring
Depth (English)2 in.
TypeSieve Cover
Diameter (English) Outer12 in.
For Use With (Equipment)12 in. dia. Test sieve
IncludesRing
Diameter (Metric) Outer305 mm
Frame MaterialStainless Steel
Mesh MaterialStainless Steel
Unit SizeEA