1x1 image pixel for data collection
U.S. Stainless Steel Test Sieve Covers
U.S. Stainless Steel Test Sieve Covers
Actual product may vary
U.S. Stainless Steel Test Sieve Covers
Sieve cover, for U.S. standard test sieve; Stainless Steel; For 8 in. dia. (20.3cm) sieves
Have Questions?
Catalog NumberDiameter (English) Outer
FIS04-887A8 in.
Catalog number FIS04-887A
Price (SGD)
104.00
EA
Add to cart
Diameter (English) Outer:
8 in.
Price (SGD)
104.00
EA
Add to cart
Cover, Sieve; Fisherbrand; For U.S. standard test sieve; Stainless Steel; For 8 in. dia. (20.3cm) sieves
Note: This is the lid only. For use with Stainless Steel pan (Cat. No. 04-887B).
  • Stainless Steel
  • Fits all 8 and 12 in. diameter (20.3 and 30.5 cm) sieves.
Specifications
TypeSieve Cover
Diameter (English) Outer8 in.
For Use With (Equipment)8 in. dia. Test sieve receiver (04-887B)
Diameter (Metric) Outer203 mm
Frame MaterialStainless Steel
Mesh MaterialStainless Steel
Unit SizeEA
Share catalog number, name or link