U.S. Standard Brass Test Sieve Covers
U.S. Standard Brass Test Sieve Covers
Actual product may vary
U.S. Standard Brass Test Sieve Covers
Cover, Sieve; Fisherbrand; For U.S. standard test sieve; Brass; For 8 in. dia. (20.3cm) sieves
Have Questions?
Catalog NumberDiameter (English) Outer
FIS04-886A8 in.
Catalog number FIS04-886A
Price (SGD)
67.00
EA
Add to cart
Diameter (English) Outer:
8 in.
Price (SGD)
67.00
EA
Add to cart
Cover, Sieve; Fisherbrand; For U.S. standard test sieve; Brass; For 8 in. dia. (20.3cm) sieves
Fit all 8 and 12 in. diameter (20.3 and 30.5cm) sieves.
Specifications
TypeSieve Cover
Diameter (English) Outer8 in.
For Use With (Equipment)8 in. dia. Test sieve
Diameter (Metric) Outer203 mm
Frame MaterialBrass
Mesh MaterialBrass
Unit SizeEA