CitriSolv™ Hybrid Solvent and Clearing Agent
Actual product may vary
CitriSolv™ Hybrid Solvent and Clearing Agent
Can be used as an alternative to xylene and ethyl acetate. CitriSolv Hybrid is soluble with absolute alcohol and will dissolve paraffin, glue, adhesives and mounting media.
Have Questions?
Catalog NumberQuantity
FIS04-355-1211 gal.
Catalog number FIS04-355-121
Price (SGD)
252.00
Each
Quantity:
1 gal.
Price (SGD)
252.00
Each
Can be used as an alternative to xylene and ethyl acetate. CitriSolv Hybrid is soluble with absolute alcohol and will dissolve paraffin, glue, adhesives and mounting media.
  • A mixture of d-limonene and hydrocarbon material
  • Reduced citrus aroma
  • Increased flash point
  • Alternative to xylene
Specifications
Chemical Name or MaterialCitriSolv™Hybrid Solvent and Clearing Agent
Physical FormLiquid
CAS5989-27-5
CAS25321-09-9
CAS128-37-0
ColorClear
Quantity1 gal.
Molecular Weight (g/mol)46.07 g/mol
Percent Purity70%
GradeLaboratory
Unit SizeEach