Contrad™ 70 Liquid Detergent
Actual product may vary
Contrad™ 70 Liquid Detergent
Cleans glass, plastic, ceramic, and ferrous metals by soaking, scrubbing, or ultrasonic cleaning
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (English)
FIS04-35534 oz.
FIS04-355-25.25 gal.
FIS04-355-355.5 gal.
Catalog number FIS04-355
Price (SGD)
80.00
Each
Volume (English):
34 oz.
Price (SGD)
80.00
Each
Cleans glass, plastic, ceramic, and ferrous metals by soaking, scrubbing, or ultrasonic cleaning
Can safely replace acid washes and caustic baths. Excellent substitute for chromic acid.
  • Concentrated liquid
  • Removes proteins, grease, oil, and baked-on contaminants
  • Nontoxic, phosphate-free, chlorine-free, and completely free-rinsing
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Specifications
Product TypeLiquid Detergent
Color-codedNo
FormConcentrated Liquid
pH13
ScentRegular Scent
SterilityNon-sterile
AppearanceWhite
Boiling Point100°C
Certifications/ComplianceUSDA, 29 CFR 1910.1200
Size1 L
Volume (English)34 oz.
Unit SizeEach