Kimble™ KIMAX™ Automatic Burets with Bottles
Kimble™ KIMAX™ Automatic Burets with Bottles
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Automatic Burets with Bottles
Class A
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-848-2C50 mL
Catalog number FIS03-848-2C
Price (SGD)
3,333.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
3,333.00
EA
Add to cart
standard taper joints fit into reagent bottles with standard taper necks.
  • Constructed with overflow tubes that return directly to reservoir
  • For air intake, overflow cups have hose connection for attaching drying tube; second drying tube provided for use between rubber bulb and reservoir
  • Precision bore tubing permanently marked with individual serial number
  • Durable black ceramic enamel scale
  • Inset shows top configuration
Specifications
Capacity (Metric)50 mL
Length (Metric)1045 mm
MaterialBorosilicate Glass
Graduations0.1 mL
For Use With (Application)Volumetric Analysis
Length (English)41.1 in.
IncludesReservoir Bottle, U-Shaped Drying Tube, Vented Connecting Tube, Rubber Squeeze Bulb, No. 1 Single-Holed Rubber Stopper, PTFE Stopcock Plug and 1/4 In. I.D. Rubber Tubing, Durable Black Ceramic Enamel Scale
ScaleBlack
Standard Taper Joint34
Stopcock MaterialPTFE
Tolerance±0.05 mL
Volume (Metric) Reservoir2000 mL
TypeAutomatic Buret
ClassClass A
Certifications/ComplianceASTM E438
Unit SizeEA