Automatic Burets with Stopcock
Automatic Burets with Stopcock
Actual product may vary
Automatic Burets with Stopcock
Ideal for storing standard solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS03-842-2B25 mL
FIS03-842-1A10 mL
Catalog number FIS03-842-2B
Price (SGD)
-
Capacity (Metric):
25 mL
  • Inset shows top configuration
  • Buret capacity: 10 and 25mL
Specifications
ColorAmber
Capacity (Metric)25 mL
MaterialGlass
Graduations0.1 mL
For Use With (Application)Storing Standard Solutions
IncludesRubber Stopper, Amber Glass Reservoir Bottle, Glass and Rubber Tubing Connections, Glass Stopcock, Rubber Bulb
ScaleAmber
Volume (Metric) Reservoir500 mL
TypeAutomatic Buret
Unit SizeEA