Polypropylene Test Tube Racks
Actual product may vary
BrandTech
Polypropylene Test Tube Racks
Constructed of sturdy polypropylene. Test Tube Racks include alphanumeric positions for easy sample identification. Racks are stackable, non-conductive, and will not float in water baths.
Have Questions?
Catalog NumberColorHolds
FIS03-840-246WhiteTubes (30 mm)
Catalog number FIS03-840-246
Price (SGD)
321.00
Each
Color:
White
Holds:
Tubes (30 mm)
Price (SGD)
321.00
Each
Sturdy polypropylene racks include alphanumeric positions for easy sample identification.
  • Racks are stackable, non-conductive and will not float in water baths.
  • Suitable for use in temperatures from -20° C to 90° C.
  • Supplied in three pieces for easy and permanent assembly.
  • Wide spaces between positions
Specifications
AutoclavableNot Autoclavable
ColorWhite
HoldsTubes (30 mm)
MaterialPolypropylene
Positions21
ShapeRectangular
StackableYes, When Empty
SterilityNon-sterile
Dimensions (L x W x H)265 x 126 x 88 mm
Length (Metric)265 mm
Length (English)10.43 in.
Width (English)4.96 in.
Height (English)3.46 in.
Diameter (Metric) Opening30 mm
Product TypeTest Tube Rack
ShelvesNone
Height (Metric)88 mm
Array3 x 7
Width (Metric)126 mm
Unit SizeEach