Nalgene™ Break-Resistant Self-Zeroing Automatic Burets
Nalgene™ Break-Resistant Self-Zeroing Automatic Burets
Actual product may vary
Nalgene™ Break-Resistant Self-Zeroing Automatic Burets
Nalgene™ Break-Resistant Self-Zeroing Automatic Burets
Nalgene™ Break-Resistant Self-Zeroing Automatic Burets
Nalgene™ Break-Resistant Self-Zeroing Automatic Burets
Thermo Scientific
Nalgene™ Break-Resistant Self-Zeroing Automatic Burets
Ensure repeatable, accurate results with these break-resistant, self-zeroing, automatic burets which meet ASTM E287.
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-834C50 mL
Catalog number FIS03-834C
Price (SGD)
623.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
623.00
EA
Add to cart
Ensure repeatable, accurate results with Thermo Scientific™ Nalgene™ Break-Resistant Self-Zeroing Automatic Burets which meet ASTM E287 standards. These Acrylic burets are crystal clear, durable, easy to use and safer than glass. Avoid chip
Features:
  • Individually calibrated to meet accuracy of ASTM E287 for repeatable, accurate results
  • Can be used with dilute mineral acids and bases except hydrofluoric acid and ammonium hydroxide
  • pH range of 1-14 is ideal; compatible with up to 5% wt/wt acids and up to 30% bases accomodating a wide range of tirants
Includes:
  • Acrylic buret
  • LDPE squeeze bottle assembly
  • 3′ (91cm) PVC tubing
  • Buret cover
Not for use with alcohols and organic solvents. Exposure to these solutions will cause immediate damage.
Specifications
ColorClear
Capacity (Metric)50 mL
Length (Metric)71.9 cm
MaterialAcrylic
Graduations0.1 mL
No. per Pack1
No. per Case4
Tolerance±0.1 mL
Volume (Metric) Reservoir1000 mL
TypeAutomatic Buret
Certifications/ComplianceASTM E287
Unit SizeEA