Specifications
Diameter (English) Outer0.55 in.
Diameter (Metric) Outer14 cm
Capacity (Metric)50 mL
Height (English)29.52 in.
MaterialGlass
Graduations0.1 mL
Height (Metric)75 cm
No. per Case1/Cs.
IncludesDust Cover
Stopcock MaterialPTFE
Tolerance±0.05 mL
TypeBuret
ClassClass A
Certifications/ComplianceASTM E287, ASTM E542
Unit SizeCase