Check Order Status
Manual Student Burets
Actual product may vary
Manual Student Burets
With blue markings.
Have Questions?
Catalog Number
Capacity (Metric)
FIS03-700B
25 mL
Catalog number
FIS03-700B
Price (SGD)
-
Capacity (Metric):
25 mL
Well suited for both educational and routine laboratory work
Specifications
Capacity (Metric)
25 mL
Material
Glass
Graduations
0.06 mL
For Use With (Application)
Educational and Routine Laboratory Work
Includes
Straight Stopcock, Blue Markings
Scale
Blue
Stopcock
Straight
Stopcock Material
Glass
Tolerance
±0.06 mL
Type
Educational Grade Buret