Kimble™ KIMAX™ Precision Bore Burets with PTFE Stopcock
Kimble™ KIMAX™ Precision Bore Burets with PTFE Stopcock
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Precision Bore Burets with PTFE Stopcock
Class A
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS03-700-22B25 mL
FIS03-700-22C50 mL
Catalog number FIS03-700-22B
Price (SGD)
934.00
Case
Add to cart
Capacity (Metric):
25 mL
Price (SGD)
934.00
Case
Add to cart
Class A
  • Precision bore tubing
  • Durable white scale
  • Permanently marked with individual serial number
  • Sold in a Case of 1
Specifications
Capacity (Metric)25 mL
Length (Metric)614 mm
MaterialBorosilicate Glass
Graduations0.1 mL
For Use With (Application)General Purpose Titrations Requiring Traceable Volumetric Accuracy
Length (English)24.17 in.
IncludesDust Cap
ScaleWhite
StopcockStraight
Stopcock MaterialPTFE
Tolerance±0.03 mL
Volume (Metric) Reservoir0.1 mL
TypeBore Buret
ClassClass A
Certifications/ComplianceASTM E287, ASTM E438, ASTM E542, ASTM E694
Unit SizeCase