Large Pails
Actual product may vary
Large Pails
Large Pails
Bel-Art
Large Pails
Rugged pails with fitted lids/covers and steel-plated bail handles
Have Questions?
Catalog NumberCapacity (Metric)Material
FIS03-687-407.6 LPolypropylene
Catalog number FIS03-687-40
Price (SGD)
984.00
Each
Capacity (Metric):
7.6 L
Material:
Polypropylene
Price (SGD)
984.00
Each
  • Translucent white pails allow for easier viewing of fluid level
  • Polypropylene is steam autoclavable (121°C), Polyethylene is not autoclavable
  • 8-qt./7.6L pail has removable steel-plated support ring and bail handle
  • 14-qt./13.2L pail has steel-plated swinging bail handle with grip, no support ring
  • Large pails meets US Specification MIL-P36257A
Specifications
Product TypePail
Capacity (Metric)7.6 L
GraduationsNone
MaterialPolypropylene
AutoclavableAutoclavable
Capacity (English)8 qt.
ColorWhite
Description8 qt. Pail
Handle MaterialPlated steel
Height (English)10 in.
Height (Metric)25 cm
IncludesRemovable support ring and steel plated bail handle, snug fitting cover
Unit SizeEach