Babcock Cream Bottles Brush
Babcock Cream Bottles Brush
Actual product may vary
Babcock Cream Bottles Brush
Fan-style tip for cleaning Babcock bottles.
Have Questions?
Catalog NumberColor
FIS03-640Black
Catalog number FIS03-640
Price (SGD)
15.00
EA
Add to cart
Color:
Black
Price (SGD)
15.00
EA
Add to cart
Fan-style tip for cleaning Babcock bottles.
  • Bristle portion on tip, 1.75 dia. × 2 in.L (4.4 × 5cm)
  • Upper portion, 0.5 dia. × 3 in.L (1.3 × 7.6cm)
Specifications
Product TypeBabcock Brush
Tip TypeFan Tip
ColorBlack
For Use With (Application)For cleaning babcock bottles
IncludesMetal ferrule
Length (English)11.5 in.
Length (Metric)29.2 cm
SizeSmall
Unit SizeEA