Transfer Pipet Brush
Transfer Pipet Brush
Actual product may vary
Fisherbrand
Transfer Pipet Brush
Cone-shaped brush with stiff nylon bristles for cleaning transfer pipets.
Have Questions?
Catalog NumberLength (English)
FIS03-62517 in.
Catalog number FIS03-625
Price (SGD)
13.00
EA
Add to cart
Length (English):
17 in.
Price (SGD)
13.00
EA
Add to cart
Clean transfer pipets with this cone-shaped brush.
  • Nylon Bristles 17 in. (43cm)
Specifications
Product TypePipet Brush
Bristle MaterialNylon
Tip TypeConical
ColorNatural
Diameter (English) Bristle0.93 in.
Diameter (Metric) Bristle2.4 cm
For Use With (Application)For cleaning transfer pipets
Length (English)17 in.
Length (English) Bristle7.5 in.
Length (Metric)43.18 cm
Length (Metric) Bristle19 cm
SizeMedium
Unit SizeEA