Nylon Tufted Brush
Nylon Tufted Brush
Actual product may vary
Fisherbrand
Nylon Tufted Brush
Unbleached bristles with tuft for test tubes.
Have Questions?
Catalog NumberColor
FIS03-590White
Catalog number FIS03-590
Price (SGD)
123.00
PK
Add to cart
Color:
White
Price (SGD)
123.00
PK
Add to cart
Unbleached bristles with tuft for test tubes.
  • Unbleached white hog bristles
  • Black, nylon tuft, 1 in. (2.5cm)
  • Galvanized wire handle
Specifications
Product TypeTest Tube Brush
Bristle MaterialNylon
Bristle TypeTufted End
Handle MaterialGalvanized Wire
Tip TypeTuft End
ColorWhite
Diameter (English) Bristle1.37 in.
Diameter (Metric) Bristle3.47 cm
For Use With (Application)For cleaning test tubes
Length (English)10 in.
Length (English) Bristle3 in.
Length (Metric)25.4 cm
Length (Metric) Bristle7.6 cm
MaterialNylon
SizeMedium
Unit SizePK