Check Order Status
Steel Wire Handled Brushes
Actual product may vary
Fisherbrand
Steel Wire Handled Brushes
Fisherbrand™ Steel Wire Handled Brushes feature stiff, white nylon bristles for cleaning test tubes.
Have Questions?
Change view
Catalog Number
Length (Metric)
Length (Metric) Bristle
Diameter (Metric) Bristle
FIS03-578
23 cm
8.9 cm
1.9 cm
FIS03-572-5
27.94 cm
10 cm
3.5 cm
2 Options
Catalog number
FIS03-578
Price (SGD)
227.00
PK
Add to cart
Length (Metric):
23 cm
Length (Metric) Bristle:
8.9 cm
Diameter (Metric) Bristle:
1.9 cm
Price (SGD)
227.00
PK
Add to cart
Fisherbrand™ Stainless-Steel Handled Brushes feature stiff, white nylon bristles for cleaning test tubes.
Bristles are chemical-resistant
Radial tufted end
Galvanized steel wire handle is corrosion resistant
Specifications
Product Type
Test Tube Brush
Bristle Material
Nylon
Bristle Type
Radial, Tufted End
Handle Material
Galvanized steel wire
Tip Type
Radial Tufted End
Color
White
Diameter (English) Bristle
0.75 in.
Diameter (Metric) Bristle
1.9 cm
For Use With (Application)
For cleaning test tubes
Length (English)
9 in.
Length (English) Bristle
3.5 in.
Length (Metric)
23 cm
Length (Metric) Bristle
8.9 cm
Size
Small