▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Brushes
▸
Budget-Priced Brushes
Actual product may vary
Fisherbrand
Budget-Priced Brushes
Fisherbrand™ Budget-Priced Brushes are an economical option for cleaning test tubes.
Have Questions?
Change view
Catalog Number
Length (Metric)
Bristle Material
Size
FIS03-573
26.67 cm
Nylon, Unbleached Hog Bristle
Large
FIS03-574
24.13 cm
Unbleached Hog Hair
Medium
FIS03-576
23 cm
Nylon
Small
3 Options
Catalog number
FIS03-573
Price (SGD)
4.00
Each
Add to cart
Length (Metric):
26.67 cm
Bristle Material:
Nylon, Unbleached Hog Bristle
Size:
Large
Price (SGD)
4.00
Each
Add to cart
Fisherbrand™ Budget-Priced Brushes are an economical option for cleaning test tubes.
03-576 is nylon with radial tufted end; rest are unbleached white hog bristles
Galvanized wire handles resist rust and corrosion
Specifications
Product Type
Test Tube Brush
Bristle Material
Nylon, Unbleached Hog Bristle
Handle Material
Galvanized Wire
Tip Type
Radial Tufted End
Color
Brown
Diameter (English) Bristle
1.37 in.
Diameter (Metric) Bristle
3.5 cm
For Use With (Equipment)
Test tubes
Length (English)
10.5 in.
Length (English) Bristle
4 in.
Length (Metric)
26.67 cm
Length (Metric) Bristle
10.16 cm
Size
Large