1x1 image pixel for data collection
Adaptable-to-Container Brush
Adaptable-to-Container Brush
Actual product may vary
Fisherbrand
Adaptable-to-Container Brush
Useful for cleaning a wide variety of glassware.
Have Questions?
Catalog NumberColor
FIS03-565Black
Catalog number FIS03-565
Price (SGD)
26.00
EA
Add to cart
Color:
Black
Price (SGD)
26.00
EA
Add to cart
12 in. brush cleans a wide variety of glassware.
  • Single-row bristles
  • Attached to heavy wire handle by swivel joint
  • Handle with ring for hanging
Specifications
Product TypeAdaptable-to-Container Brush
Handle MaterialHeavy Wire
ColorBlack
Diameter (English) Bristle1.5 in.
Diameter (Metric) Bristle3.8 cm
For Use With (Application)For cleaning a wide variety of glassware
IncludesRing
Length (English)12 in.
Length (English) Bristle4.25 in.
Length (Metric)30.5 cm
Length (Metric) Bristle10.8 cm
No. of Bristle Rows1
SizeMedium
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link