Reusable Aluminum Sample Container
Reusable Aluminum Sample Container
Actual product may vary
Reusable Aluminum Sample Container
Excellent for moisture determinations in soil analysis
Have Questions?
Catalog NumberNo. per Pack
FIS03-485C12/Pk.
Catalog number FIS03-485C
Price (SGD)
74.00
EA
Add to cart
No. per Pack:
12/Pk.
Price (SGD)
74.00
EA
Add to cart
Collect soil in this container, weigh it, then dry it in oven. Moisture content is determined by reweighing soil after drying.
  • Round seamless aluminum
  • Recommended for general use
Specifications
MaterialAluminum
Certifications/ComplianceASTM™
Diameter (Metric)8.9 cm
Height (English)2 in.
Height (Metric)5 cm
No. per Case144/Cs.
No. per Pack12/Pk.
Unit SizeEA