Sterilizer Box for Petri Dishes
Sterilizer Box for Petri Dishes
Actual product may vary
Fisherbrand
Sterilizer Box for Petri Dishes
Fisherbrand Sterilizer Box holds and protects petri dishes during and after sterilization.
Have Questions?
Catalog NumberHolds
FIS03-46010 (3.93 x 0.59 in. Glass Petri Dishes), 18 (3.93 x 0.39 in. Glass Petri Dishes)
Catalog number FIS03-460
Price (SGD)
201.00
EA
Add to cart
Holds:
10 (3.93 x 0.59 in. Glass Petri Dishes), 18 (3.93 x 0.39 in. Glass Petri Dishes)
Price (SGD)
201.00
EA
Add to cart
  • Fisherbrand™ Sterilizer Box holds and protects petri dishes during heat sterilization with deep telescoping cover and removable rack.
    • Polished stainless-steel, including deep telescoping cover and removable rack
    • Holds 18 glass Petri dishes size 3.93 x 0.39 in. (100 x 10mm), or 10 size 3.93 x 0.59 in. (100 x 15mm)
    • Overall, 4.25 O.D. x 10.50 in.H (12 x 27cm)
Specifications
MaterialStainless Steel
ShapeRound
For Use With (Application)Protects Petri Dishes During and After Heat Sterilization
Holds10 (3.93 x 0.59 in. Glass Petri Dishes), 18 (3.93 x 0.39 in. Glass Petri Dishes)
IncludesDeep Telescoping Cover, Removable Rack
Product TypeSterilization Box
Height (Metric)27 cm
Height (English)10.5 in.
Diameter (English) Outer4.25 in.
Diameter (Metric) Outer12 cm
Unit SizeEA