WHEATON™ LDPE Dropping Bottles with Extended Controlled Dropper Tips and Polypropylene Dropper Tip Caps
Actual product may vary
WHEATON™ LDPE Dropping Bottles with Extended Controlled Dropper Tips and Polypropylene Dropper Tip Caps
WHEATON™ LDPE Dropping Bottles with Extended Controlled Dropper Tips and Polypropylene Dropper Tip Caps
DWK Life Sciences
WHEATON™ LDPE Dropping Bottles with Extended Controlled Dropper Tips and Polypropylene Dropper Tip Caps
WHEATON™ LDPE Dropping Bottles with Extended Controlled Dropper Tips and Polypropylene Dropper Tip Caps deliver precise, repeatable dispensing — one drop at a time. Average drop size is 44μL ±6μL. Radiation-grade polypropylene caps and tips are packaged separately.
Have Questions?
Catalog NumberCapacity (Metric)Color
FIS03-439-61P7 mLWhite
Catalog number FIS03-439-61P
Price (SGD)
749.00
Case
Capacity (Metric):
7 mL
Color:
White
Price (SGD)
749.00
Case
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size: 44μL ±6μL
  • Bottle made of LDPE (Low Density Polyethylene)
  • Radiation-grade polypropylene screw caps and tips packaged separately
  • Available with natural or white dropper tips
  • Bottles, tips and caps are packaged separately
Specifications
Capacity0.24 oz., 7 mL
Capacity (Metric)7 mL
ColorWhite
Diameter (English)0.74 in.
IncludesExtended controlled dropper tip
MaterialLDPE
SterilityNon-sterile
Closure IncludedYes
GraduatedNo
MouthNarrow
Height (Metric)63 mm
Height (English)2.48 in.
Closure MaterialPolypropylene
Closure Size15-415 mm
Diameter (Metric)19 mm
AutoclavableNot Autoclavable
Capacity (English)0.24 oz.
ShapeRound
DisposableReusable
Product TypeDispensing Bottle
Unit SizeCase