Specifications
Mesh MaterialPolyethylene
ColorWhite
For Use With (Application)Provide fast and efficient filtering of small volume samples (up to 1000μL) prior to FLOW or FACS™ analysis
IncludesCompact tray
Product TypeCell Strainer
SterilitySterile
For Use With (Equipment)Pipette Tips, 1000 μL, including Fisherbrand Sure-One, VWR, Axygen, Nichiryo, Eppendorf and more
Mesh Size70 μm
Unit SizePack