▸
Pumps
▸
Drum Pumps
▸
All-Polyethylene Poly-Hand Pumps
Actual product may vary
Bel-Art
All-Polyethylene Poly-Hand Pumps
Compatible with all standard drum bungs or carboys
Have Questions?
Catalog Number
For Use With
FIS03-420-125
5 gal. Drums
Catalog number
FIS03-420-125
Price (SGD)
1,233.00
Each
Add to cart
For Use With:
5 gal. Drums
Price (SGD)
1,233.00
Each
Add to cart
Threads directly to standard drum bung or carboy
Constructed entirely of rugged polyethylene
Inert to strong acids, alkalis and many solvents
Delivers by positive pressure on the downstroke of the piston; does not pressurize the vessel
Ergonomic handle fits comfortably in gloved hand
Pump will efficiently deliver against 3.65 m (12 ft.) head of water and empty drum or carboy to within 3 mm of the bottom
Sizes are available to fit 114 and 220L (30-50 gal) plastic or steel drums and 20L (5 gal) carboy
Specifications
Applications
Industrial liquid handling
Flow Rate
560 mL/stroke
For Use With
5 gal. Drums
Pump Tube Length
For 20L Carboys
Resistance Properties
Alkalis, Detergent, Acid
Type
Drum Pump
Material
Polyethylene