Cryo-Safe™ Microcentrifuge Tube and Cryogenic Vial Coolers
Cryo-Safe™ Microcentrifuge Tube and Cryogenic Vial Coolers
Actual product may vary
Bel-Art
Cryo-Safe™ Microcentrifuge Tube and Cryogenic Vial Coolers
Benchtop-sized coolers for microcentrifuge tubes or cryogenic vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHoldsDimensions (L x W x H)Weight (Metric)
FIS03-410-501White, Yellow32 Tubes (0.5 to 2 mL)9.5 x 3.19 x 5.7 in.2 kg
FIS03-410-500Gray, Red32 Tubes (0.5 to 1.5 mL)9.5 x 3.19 x 5.7 in.2 kg
Catalog number FIS03-410-501
Price (SGD)
885.00
EA
Add to cart
Color:
White, Yellow
Holds:
32 Tubes (0.5 to 2 mL)
Dimensions (L x W x H):
9.5 x 3.19 x 5.7 in.
Weight (Metric):
2 kg
Price (SGD)
885.00
EA
Add to cart
Benchtop-sized coolers for microcentrifuge tubes or cryogenic vials.
  • Holds Twelve 0.5 or 1.5mL tube in individual wells below 0 deg.C for 2.5 Hours
  • Body of cooler filled with a nontoxic gel that does not contact the contents of the cooler
  • Excellent for safe transport of temperature sensitive tubes
  • Polycarbonate body and lid is easy to clean
  • Stackable design with rubber feet to prevent sliding
  • Mini Coolers feature molded-in side handles, Junior and Maxi Coolers have fold down wire carrying handle that helps retain lid
  • Store -15°C coolers at -15°C for 24 hours prior to use
  • Store 0°C coolers at -20°C for a few hours prior to use
  • Store -20°C coolers at -20°C for 24 hours prior to use
Specifications
Height (Metric)14.6 cm
Height (English)5.7 in.
Length (Metric)24.3 cm
Length (English)9.5 in.
Width (Metric)15.7 cm
Width (English)3.19 in.
Dimensions (L x W x H)9.5 x 3.19 x 5.7 in.
Weight (English)4.5 lb.
Weight (Metric)2 kg
MaterialPolycarbonate
TypeBenchtop Cooler
ColorWhite, Yellow
Capacity (Metric)32 tubes
Holds32 Tubes (0.5 to 2 mL)
AutoclavableAutoclavable
Temperature ControlNo
Temperature Range-20°C
Unit SizeEA