Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Size |
|---|---|---|
| FIS03-409-10AA | 125 mL | 24 mm |
| 2401-0125 | 125 mL | 24 mm |
| 2401-0250 | 250 mL | 24 mm |
| FIS03-409-10BB | 250 mL | 24 mm |
| FIS03-409-10CC | 500 mL | 28 mm |
| 2401-1000 | 1000 mL | 38-430 mm |
| FIS03-409-10DD | 1000 mL | 38-430 mm |
| FKS10048300 | 1000 mL | 38-430 mm |
High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
Features:
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.