LDPE Dispensing Bottles
LDPE Dispensing Bottles
Actual product may vary
LDPE Dispensing Bottles
Versatile, reliable bottles and spouts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FIS03-402G500 mL28-400 mm
FIS03-402A15 mL15-415 mm
FIS03-402B30 mL18 mm
FIS03-402D125 mL24-400 mm
Catalog number FIS03-402G
Price (SGD)
98.00
PK
Add to cart
Capacity (Metric):
500 mL
Closure Size:
28-400 mm
Price (SGD)
98.00
PK
Add to cart
Versatile, reliable bottles and spouts.
  • LDPE bottles and spouts with sealer caps
  • Sealer snaps on easily after use and gives tight seal
  • Spout openings are standard 0.03 in. (0.08cm)
  • Can be enlarged by cutting back with scissors
Specifications
Capacity16 oz., 500mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
MaterialLDPE
Width (Metric)0.12mm
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)0.3 mm
Product TypeDispensing Bottle
Width (English)3.07 in.
Height (English)7.75 in.
Closure MaterialLDPE
Closure Size28-400 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePK