Polypropylene 15 and 50 mL Vial Storage Boxes
Actual product may vary
Polypropylene 15 and 50 mL Vial Storage Boxes
Accommodate 15- or 50-mL centrifuge tubes for long-term storage
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsColorDesign
FIS03-395-64736 TubesBlue15mL box
FIS03-395-64816 TubesOrange50mL box
Catalog number FIS03-395-647
Price (SGD)
131.00
Pack
Holds:
36 Tubes
Color:
Blue
Design:
15mL box
Price (SGD)
131.00
Pack
Accommodates centrifuge tubes for long-term storage.
  • Store down to -90°C (-130°F) or autoclave up to 122°C (252°F)—wide temperature range
  • Inspect, reference, and identify samples through the and clear lid and easy-to-read numeric grid
  • Label or mark boxes on the white top portion
  • Compatible with most 4 in. stainless steel freezer racks
These cryogenic storage boxes offer excellent low-temperature storage. Polypropylene boxes have clear lids with colored inserts and numbered grids to facilitate easy sample identification.
Specifications
ColorBlue
For Use With (Application)Long-term centrifuge tubes storage
For Use With (Equipment)Centrifuge tubes
Holds36 Tubes
Design15mL box
MaterialPolypropylene
TypeFreezer Box
Withstand Temperature (Metric)-90°C
Unit SizePack