Search Fisher Scientific
| Catalog Number | Dimensions (L x W x H) | Includes |
|---|---|---|
| FIS03-395-455 | 5 x 5 x 2 in. | Bottom Drain Holes, Tight Lid, No Divider |
| FIS03-395-454 | 5.75 x 5.75 x 4.8 in. | Bottom Drain Holes, Tight Lid, No Divider |
| FIS03-395-465 | 5 x 5 x 2 in. | Bottom Drain Holes, Tight Lid, 100-Cell Divider |
| FIS03-395-463 | 3 x 3 x 2 in. | Bottom Drain Holes, Tight Lid, 25-Cell Divider |
| FIS03-395-464 | 5 x 5 x 2 in. | Bottom Drain Holes, Tight Lid, 81-Cell Divider |
Choice of boxes to accommodate tubes from 0.5 to 50mL
Sturdy yet economical, Fisherbrand Cryo/Freezer Boxes and Dividers are made of sturdy white cardboard, with a special moisture-repellent coating to protect them from the effects of liquid nitrogen or condensation from refrigerated equipment. Small holes on the bottom allow for safe drainage of liquid nitrogen or water.
Simple design for versatile use
Choice of sizes and dividers for flexibility
Provide low-cost storage for laboratories in universities, government, medical research, and pharmaceutical facilities; In institutions performing artificial insemination, biotech, blood bank, and stem cell/bone marrow procedures.