Cryo/Freezer Boxes
Actual product may vary
Cryo/Freezer Boxes
Choice of boxes to accommodate tubes from 0.5 to 50mL
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)Includes
FIS03-395-4545.75 x 5.75 x 4.8 in.Bottom Drain Holes, Tight Lid, No Divider
FIS03-395-4555 x 5 x 2 in.Bottom Drain Holes, Tight Lid, No Divider
FIS03-395-4655 x 5 x 2 in.Bottom Drain Holes, Tight Lid, 100-Cell Divider
FIS03-395-4633 x 3 x 2 in.Bottom Drain Holes, Tight Lid, 25-Cell Divider
FIS03-395-4645 x 5 x 2 in.Bottom Drain Holes, Tight Lid, 81-Cell Divider
Catalog number FIS03-395-454
Price (SGD)
12.00
Each
Dimensions (L x W x H):
5.75 x 5.75 x 4.8 in.
Includes:
Bottom Drain Holes, Tight Lid, No Divider
Price (SGD)
12.00
Each
Choice of boxes to accommodate tubes from 0.5 to 50mL
Sturdy yet economical, Fisherbrand Cryo/Freezer Boxes and Dividers are made of sturdy white cardboard, with a special moisture-repellent coating to protect them from the effects of liquid nitrogen or condensation from refrigerated equipment. Small holes on the bottom allow for safe drainage of liquid nitrogen or water.
Simple design for versatile use
  • Unbreakable white fiberboard to protect samples
  • Surface is easy to write on for identification
  • Sized to fit all standard stainless-steel and polycarbonate storage systems
  • Protective coating prevents water or liquid nitrogen from saturating box for increased longevity
  • Tight lid-to-box fit means accidental separation is next to impossible
  • Bottom drain holes allow safe drainage of liquid nitrogen, prevent moisture buildup
  • Catalog numbers are printed on box lids for easy reorder
  • Can be used in mechanical freezers and liquid nitrogen tanks at temperatures to -196°C
Choice of sizes and dividers for flexibility
  • Three boxes come with dividers in popular configurations
  • Four boxes come without dividers: choose the divider you need, replace it with another divider when needs change
Provide low-cost storage for laboratories in universities, government, medical research, and pharmaceutical facilities; In institutions performing artificial insemination, biotech, blood bank, and stem cell/bone marrow procedures.
Specifications
ColorWhite
MaterialFiberboard
TypeCryo/Freezer Box
For Use With (Application)Provides low-cost storage for laboratories in universities, government, medical research, and pharmaceutical facilities; in institutions performing artificial insemination, biotech, blood bank, and stem cell/bone marrow procedures.
For Use With (Equipment)Used in mechanical freezers and liquid nitrogen tanks and all standard stainless steel and polycarbonate storage systems.
IncludesBottom Drain Holes, Tight Lid, No Divider
Max. Temperature (English)-320.8°F
Withstand Temperature (Metric)-196°C
Dimensions (L x W x H)5.75 x 5.75 x 4.8 in.
Unit SizeEach