Dividers for Cryo/Freezer Boxes
Dividers for Cryo/Freezer Boxes
Actual product may vary
Dividers for Cryo/Freezer Boxes
For Fisherbrand Cryo/Freezer Boxes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsLength (Metric)Width (Metric)Height (Metric)
FIS03-395-4522512.19 cm12.3 cm3.3 cm
FIS03-395-45710012.3 cm12.3 cm3.3 cm
FIS03-395-4588112.19 cm12.3 cm3.3 cm
FIS03-395-4611614.2 cm14.22 cm6.9 cm
FIS03-395-4623614.2 cm14.19 cm6.9 cm
Catalog number FIS03-395-452
Price (SGD)
4.00
Each
Add to cart
Holds:
25
Length (Metric):
12.19 cm
Width (Metric):
12.3 cm
Height (Metric):
3.3 cm
Price (SGD)
4.00
Each
Add to cart
For Fisherbrand Cryo/Freezer Boxes
These Fisherbrand box dividers are made with unbreakable white fiberboard to safeguard samples. A protective coating prevents water or liquid nitrogen from saturating box for increased longevity. Can be used in mechanical freezers and liquid nitrogen tanks at temperatures to -196°C (-320.8°F).
  • Nine divider choices hold from 16 to 144 tubes
  • Accommodate tubes with diameters from 9.8 to 33.5mm
Provide low-cost storage for laboratories in universities, government, medical research, and pharmaceutical facilities; in institutions performing artificial insemination, biotech, blood bank, and stem cell/bone marrow procedures.
Specifications
MaterialFiberboard
Product TypeBox Divider
For Use With (Application)Biotech, Blood Bank, Government, Institutions Performing Artificial Insemination, Medical Research and Pharmaceutical Facilities, Provides Low-Cost Storage for Laboratories in Universities, Stem Cell/Bone Marrow Procedures
Holds25
Height (Metric)3.3 cm
ColorWhite
For Use With (Equipment)Fisherbrand Cryo/Freezer Boxes, Mechanical Freezers and Liquid Nitrogen Tanks
Width (Metric)12.3 cm
Length (Metric)12.19 cm
ShapeSquare
Height (English)1.3 in.
Length (English)4.8 in.
Width (English)4.8 in.
Unit SizeEach