Storage Vials
Storage Vials
Actual product may vary
Thermo Scientific
Storage Vials
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorClosure SizeQuantity
FIS03-394EWhite, Clear100
FIS03-391-10A2 mLClear, Clear8-425100 Vials
FIS03-391-10B4 mLClear100
FIS03-391-10D12 mLClear200
FIS03-391-10F22 mLClear200 (vial and caps are sold separately)
FIS03-391-11B4 mLAmber100
FIS22-0456818 mLAmber200
FKS1249867612 mLAmber200
FIS03-394AWhite, Clear15-425100 Septa
FIS03-394CWhite, Clear100
FIS10-800-145White15 mm100
Catalog number FIS03-394E
Price (SGD)
-
Color:
White, Clear
Quantity:
100
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSepta for 8-425 Screw Cap
ColorWhite, Clear
MaterialPTFE, Silicone
Quantity100
Thickness (Metric)1.5 mm
TypeSeptum Only
Unit SizePK