Acrylic Safety Glasses Holder 12-Unit
Acrylic Safety Glasses Holder 12-Unit
Actual product may vary
Fisherbrand
Acrylic Safety Glasses Holder 12-Unit
Clean line storage solutions perfect for laboratory organization. Fisherbrand Acrylic Safety Glasses Holder 12-Unit is made of clear durable acrylic.
Have Questions?
Catalog number FIS03-391-578
Price (SGD)
257.00
EA
Add to cart
Price (SGD)
257.00
EA
Add to cart
Clean line storage solutions perfect for laboratory organization. Fisherbrand Acrylic Safety Glasses Holder 12-Unit is made of clear durable acrylic.
  • Built-in handles on either side allow for easy setup
  • Holder can be mounted vertically, horizontally or placed on the benchtop
  • Holder offers versatility while providing space-saving storage
  • All mounting accessories included
Specifications
Height (English)9.25 in.
Height (Metric)23.5 cm
Width (English)6.75 in.
Width (Metric)17.145 cm
ColorClear
Device Holds12 safety glasses
Product TypeEyewear Storage Box
For Use With (Application)Storing 12 pairs of safety glasses
IncludesMounting accessories
Length (English)12.25 in.
Length (Metric)31.11 cm
MaterialAcrylic