Acrylic Paper Towel Dispenser
Actual product may vary
Acrylic Paper Towel Dispenser
Clean line storage solutions perfect for laboratory organization. Fisherbrand Acrylic Paper Towel Dispenser is made of clear durable acrylic.
Have Questions?
Catalog NumberMounting
FIS03-391-572Wall or Surface Mount
Catalog number FIS03-391-572
Price (SGD)
158.00
Each
Mounting:
Wall or Surface Mount
Price (SGD)
158.00
Each
  • Simply designed to accommodate a variety of C-fold or Z-fold paper towels
  • Can be wall mounted or placed directly on the benchtop
  • Holder offers versatility while providing space-saving storage
  • All mounting accessories included.
Specifications
Product TypePaper Towel Dispenser
ColorClear
Length (English)10.5 in.
Length (Metric)26.67 cm
Width (English)6.38 in.
Width (Metric)16.2 cm
HoldsPaper Towels
MaterialAcrylic
MountingWall or Surface Mount
No. of Compartments1
Product StyleOpen Top and Front
For Use With (Application)Holding paper towels
IncludesMounting accessories
Unit SizeEach