11mm Autosampler Vial Crimp Caps, Kombo Kit; 200 of Each Color; Silver, Blue, Green, Red, Yellow Aluminum Seals; Seal 2000 PTFE/Rubber Septa
11mm Autosampler Vial Crimp Caps, Kombo Kit; 200 of Each Color; Silver, Blue, Green, Red, Yellow Aluminum Seals; Seal 2000 PTFE/Rubber Septa
Actual product may vary
Thermo Scientific
11mm Autosampler Vial Crimp Caps, Kombo Kit; 200 of Each Color; Silver, Blue, Green, Red, Yellow Aluminum Seals; Seal 2000 PTFE/Rubber Septa
11mm Crimp Top Closures feature pre-assembled caps and septa to minimize contamination from handling. They are compatible with most HPLC and GC autosamplers.
Have Questions?
Catalog NumberColor
FIS03-378-347Silver, Blue, Green, Red, Yellow
Catalog number FIS03-378-347
Price (SGD)
-
Color:
Silver, Blue, Green, Red, Yellow
These crimp caps are conveniently pre-assembled caps and septa to minimize contamination from handling. They are compatible with most HPLC and GC autosamplers. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands. Aluminum Crimp Seals with prefitted septa fit all 11mm Crimp Top and Snap Cap Vials and must be applied with a crimping tool. Closures are shipped in sealed polybags to prevent contamination during transport.
  • Anodized aluminum seals fit all 11mm crimp-top vials
  • Seal 2000™ material is specially formulated for improved background performance over PTFE/red rubber
  • Viton™ septa perform well with ECD detectors and are resistant to many aggressive solvents
Specifications
DescriptionKombo Kit; 200 of Each Color; Silver, Blue, Green, Red, Yellow Aluminum Seals; Seal 2000 PTFE/Rubber Septa
ColorSilver, Blue, Green, Red, Yellow
MaterialAluminum
SeptumPTFE/Synthetic Red Rubber
Quantity1000
Thickness (Metric)1.0 mm
Closure Size11 mm
Unit SizeCS