11 mm Autosampler Vial Crimp Caps
11 mm Autosampler Vial Crimp Caps
Actual product may vary
Fisherbrand
11 mm Autosampler Vial Crimp Caps
Minimize contamination from handling with pre-assembled caps and septa; compatible with most HPLC and GC autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSeptumMaterialType
FIS03-377-91Silver, RedPTFE/Synthetic Red RubberPTFE, Rubber, AluminumCrimp Cap
FIS06-406-19BSilver, Clear, RedClear PTFE/Natural Red RubberPTFE, Rubber, AluminumCrimp Cap
Catalog number FIS03-377-91
Price (SGD)
-
Color:
Silver, Red
Septum:
PTFE/Synthetic Red Rubber
Material:
PTFE, Rubber, Aluminum
Type:
Crimp Cap
Minimize contamination from handling with convenient, pre-assembled Thermo Scientific™ 11 mm Autosampler Vial Crimp Caps, compatible with most HPLC and GC autosamplers. Choose from Thermo Scientific™, National™ or Chromacol™ brands.
Aluminum crimp seals with prefitted septa fit all 11 mm crimp top and snap cap vials. Must be applied with a crimping tool.
Ordering Information:
Shipped in sealed polybags to prevent contamination during transport.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionClear PTFE/NSC Silicone Septa; Silver Aluminum Seal
ColorSilver, Red
MaterialPTFE, Rubber, Aluminum
SeptumPTFE/Synthetic Red Rubber
Quantity100
Thickness (Metric)1.0 mm
TypeCrimp Cap
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePK