National™ Headspace 20mm Crimp Seals with Septa
Actual product may vary
Thermo Scientific
National™ Headspace 20mm Crimp Seals with Septa
National Crimp Seals ensure sample security every time.
Have Questions?
Catalog NumberColor
FIS03-377-297Amber
Catalog number FIS03-377-297
Price (SGD)
-
Color:
Amber
20mm Crimp seals with prefitted septa for headspace vials
  • Seals with pre-inserted septa are convenient and minimize contamination due to handling
  • Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
  • PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
  • Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required
SepCap is for short-term storage. Do not use SepCap at elevated temperatures.
Specifications
ColorAmber
MaterialSilica Glass
Product TypeSepta
Unit SizePack