20 mm Headspace Vials, Septum, and Caps
20 mm Headspace Vials, Septum, and Caps
Actual product may vary
Thermo Scientific
20 mm Headspace Vials, Septum, and Caps
Headspace vials are available in clear or amber glass with a round or flat base, a crimp seal with a beveled or square edge finish, or with a screw-thread finish. Headspace vials, septum, and caps can be purchased in various quantities separately or together as convenience kits to fit your laboratory needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorTypeDiameter (Metric) InnerMaterial
FIS03-375-22ESilver, Tan, WhiteCrimp CapPTFE, Silicone, Aluminum
FIS03-377-119Clear20 mm Headspace Crimp Vial23 mmGlass
FKS11905713Clear20 mm Headspace Crimp Vial23 mmGlass
FIS03-377-118Clear20 mm Headspace Crimp Vial Convenience Kit20 mmGlass
Catalog number FIS03-375-22E
Price (SGD)
-
Color:
Silver, Tan, White
Type:
Crimp Cap
Material:
PTFE, Silicone, Aluminum
Thermo Scientific™ headspace vials are available in clear or amber glass with a round or flat base, a crimp seal with a beveled or square edge finish, or with a screw-thread finish. Headspace vials, septum, and caps can be purchased in various quantities separately or together as convenience kits to fit your laboratory needs.
  • Type 1 borosilicate glass
  • Clear glass vials with 20mm crimp seal finish are designed to fit most headspace autosamplers
  • Flat bottom vials maximize heating efficiency when used with bottom
  • Round bottom vials are more easily handled by robotic arms that lift the vial from the tray
  • Round bottom vials distribute the internal pressure created at high temperatures across the glass surface
  • Beveled edge finish features a sturdy rim that presses into the septum for a more effective seal
Compatibility
  • Flat bottom vials feature a longer neck for fit with Carlo Erba™ and Agilent™ units
  • Round bottom vials are designed for Perkin Elmer™, Tekmar™ and Varian™ units
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description20mm Crimp Cap, 9.5mm hole
ColorSilver, Tan, White
MaterialPTFE, Silicone, Aluminum
SeptumTan PTFE/White Silicone
Quantity100
Thickness (Metric)3.2 mm
TypeCrimp Cap
Closure Size20 mm
For Use With (Equipment)20 mm Headspace Vials
Unit SizePK