MaterialBorosilicate Glass
Capacity (Metric)10 mL, 100 mL, 25 mL, 250 mL, 50 mL
For Use With (Application)Start Up Labs and Customers Who Need a Variety of Cylinders But Have Limited Lab Space or Glassware Needs
ScaleSingle
Graduations0.2 mL (10 mL), 0.5 mL (25 mL), 1 mL (100 mL), 1 mL (50 mL), 2 mL (250 mL)
Subdivision0.2 to 2mL
CalibrationTo Deliver
Base TypeRound
For Use With (Equipment)Bumper
ShapeRound
ClassClass B
Height (Metric)190 mm (50 mL), 220 mm (250 mL), 255 mm (100 mL), 135 mm (10 mL), 140 mm (25 mL)
Certifications/ComplianceASTM E1272, ASTM E438, USP Type I
Tolerance±0.1 mL (at 10 mL), ±0.3 mL (at 25 mL), ±0.4 mL (at 50 mL), ±0.6 mL (at 100 mL), ±1.4 mL (at 250 mL)
Product TypeGraduated Cylinder Set
Unit SizeEach