51 Expansion Short Style Glass Shell Vials without Closures
Actual product may vary
Fisherbrand
51 Expansion Short Style Glass Shell Vials without Closures
Short-style shell vials are ideal for storing dry products.
Have Questions?
Catalog NumberVolume (Metric)Height (Metric)Diameter (Metric) Outer
FIS03-339-30B2 mL35 mm12 mm
Catalog number FIS03-339-30B
Price (SGD)
59.00
Pack
Volume (Metric):
2 mL
Height (Metric):
35 mm
Diameter (Metric) Outer:
12 mm
Price (SGD)
59.00
Pack
Short-style shell vials are ideal for storing dry products.
  • Plain top design
  • Lab pack quantities
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
ColorClear
Diameter (Metric) Outer12 mm
MaterialBorosilicate Glass
SterilityNon-sterile
DisposableSingle-use
Volume (Metric)2 mL
For Use With (Application)Ideal for storing dry products
No. per Pack144
Volume (English)0.5 dr.
Volume (Metric) Total2 mL
Certifications/ComplianceASTM E438
Height (Metric)35 mm
Product TypeShell Vial
Unit SizePack