Kimble™ Disposable Glass Coliwasa Tube
Actual product may vary
DWK Life Sciences
Kimble™ Disposable Glass Coliwasa Tube
Prescored; snaps apart for disposal
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-338-2F200 mL
Catalog number FIS03-338-2F
Price (SGD)
1,252.00
Case
Capacity (Metric):
200 mL
Price (SGD)
1,252.00
Case
  • Ball in lower end of two-part borosilicate glass tube acts as a check valve to prevent sample loss during transfer
Specifications
Product TypeColiwasa Tube
MaterialGlass
Capacity (English)6.76 oz.
Capacity (Metric)200 mL
Certifications/ComplianceType I and ASTM E438, Type I, Class A
Diameter (English) Outer0.86 in.
Diameter (Metric) Outer2.2 cm
Length (English)42 in.
Length (Metric)106.6 cm
Unit SizeCase