Kimble™ Disposable Drum Sampler
Actual product may vary
DWK Life Sciences
Kimble™ Disposable Drum Sampler
Borosilicate glass, open-end drum thieves break easily for quick disposal. Available in 150mL, 25mL and 75mL capacity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (English) OuterDiameter (Metric) Outer
FIS03-338-2E150 mL0.7 in.1.8 cm
FIS03-338-2D75 mL0.47 in.1.2 cm
Catalog number FIS03-338-2E
Price (SGD)
864.00
Case
Capacity (Metric):
150 mL
Diameter (English) Outer:
0.7 in.
Diameter (Metric) Outer:
1.8 cm
Price (SGD)
864.00
Case
  • Conventional open-end drum thieves provide a quick method for sampling unmarked drums
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Product TypeDrum Sampler
MaterialBorosilicate Glass
Capacity (English)5.07 oz.
Capacity (Metric)150 mL
DisposableSingle-use
For Use With (Application)Provides a quick method for sampling unmarked drums
Diameter (English) Outer0.7 in.
Diameter (Metric) Outer1.8 cm
Length (English)43 in.
Length (Metric)109.2 cm
Unit SizeCase