▸
Environmental Samplers
▸
Liquid Samplers
▸
Kimble™ Disposable Drum Sampler
Actual product may vary
DWK Life Sciences
Kimble™ Disposable Drum Sampler
Borosilicate glass, open-end drum thieves break easily for quick disposal. Available in 150mL, 25mL and 75mL capacity.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Diameter (English) Outer
Diameter (Metric) Outer
FIS03-338-2D
75 mL
0.47 in.
1.2 cm
FIS03-338-2E
150 mL
0.7 in.
1.8 cm
2 Options
Catalog number
FIS03-338-2D
Price (SGD)
506.00
Case
Add to cart
Capacity (Metric):
75 mL
Diameter (English) Outer:
0.47 in.
Diameter (Metric) Outer:
1.2 cm
Price (SGD)
506.00
Case
Add to cart
Conventional open-end drum thieves provide a quick method for sampling unmarked drums
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Product Type
Drum Sampler
Material
Borosilicate Glass
Capacity (English)
2.5 oz.
Capacity (Metric)
75 mL
Disposable
Single-use
For Use With (Application)
Provides a quick method for sampling unmarked drums
Diameter (English) Outer
0.47 in.
Diameter (Metric) Outer
1.2 cm
Length (English)
43 in.
Length (Metric)
109.2 cm