HDPE Liquid Samplers
Actual product may vary
HDPE Liquid Samplers
For industrial and environmental applications.
Have Questions?
Catalog NumberLength (Metric) HandleLength (English) HandleWidth (Metric)Width (English)
FIS03-338-2B1.8 m6 ft.7.62 cm3 in.
Catalog number FIS03-338-2B
Price (SGD)
450.00
Each
Length (Metric) Handle:
1.8 m
Length (English) Handle:
6 ft.
Width (Metric):
7.62 cm
Width (English):
3 in.
Price (SGD)
450.00
Each
Chemical-resistant, high-density polyethylene samplers ideal for use in corrosive environments and hard-to-reach places.
  • 500mL bottle screws into the end of handle and is filled via four ports
  • Handle is supplied in two pieces
  • Reusable
  • Handle O.D: 1.25 in. (3.2cm)
Specifications
Product TypeLiquid Sampler
MaterialHDPE
For Use With (Application)For industrial and environmental applications
Diameter (English) Handle1.25 in.
Diameter (Metric) Handle3.2 cm
Height (English)81.06 in.
Height (Metric)205.89 cm
Length (English) Handle6 ft.
Length (Metric) Handle1.8 m
Width (English)3 in.
Width (Metric)7.62 cm
Unit SizeEach