Graduated HDPE Rectangular Bottles
Graduated HDPE Rectangular Bottles
Actual product may vary
Graduated HDPE Rectangular Bottles
Save valuable shelf space while providing greater materials storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS03-311-3G2000 mL
FIS03-311-3E500 mL
Catalog number FIS03-311-3G
Price (SGD)
181.00
Pack
Add to cart
Capacity (Metric):
2000 mL
Price (SGD)
181.00
Pack
Add to cart
Save valuable shelf space while providing greater materials storage.
Ideal for shipping and storing solid chemicals, liquids and pastes.
  • Graduated in mL
  • Wide mouth for convenient cleaning, sample filling and retrieval
  • HDPE screw cap with LDPE inner security seal eliminates leaks
  • Attachment holes on closure and bottle shoulder permit attachment of identification tag or custody seal
Specifications
Capacity64 oz., 2000mL
Capacity (Metric)2000 mL
Capacity (English)64 oz.
ColorWhite, Translucent
DescriptionSquare Bottle HDPE 2000mL
Diameter (English) Inner Neck2.2 in.
Diameter (Metric) Outer Neck58 mm (Inner Neck)
Diameter (Metric) Inner Neck58 mm
MaterialHDPE
Length (Metric)100mm
Max. Temperature (Metric)110°C
Max. Temperature (English)230°F
Neck ID (Metric)18mm
No. per Case12
No. per Pack6
QuantityCase, Pack
SeptaYes
SterilityNon-sterile
TransparencyTranslucent
Width (Metric)134mm
Closure ColorTranslucent
Graduations200mL
Closure IncludedYes
GraduatedYes
MouthWide
ShapeSquare
Height (Metric)210 mm
Product TypeSquare Bottle
Width (English)5.28 in.
Length (English)3.94 in.
Height (English)8.37 in.
Closure MaterialHDPE
Closure TypeScrew Cap
Through the Closure DispensingNo