Graduated HDPE Rectangular Bottles
Actual product may vary
Graduated HDPE Rectangular Bottles
Save valuable shelf space while providing greater materials storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS03-311-3E500 mL
FIS03-311-3G2000 mL
Catalog number FIS03-311-3E
Price (SGD)
59.00
Pack
Capacity (Metric):
500 mL
Price (SGD)
59.00
Pack
Save valuable shelf space while providing greater materials storage.
Ideal for shipping and storing solid chemicals, liquids and pastes.
  • Graduated in mL
  • Wide mouth for convenient cleaning, sample filling and retrieval
  • HDPE screw cap with LDPE inner security seal eliminates leaks
  • Attachment holes on closure and bottle shoulder permit attachment of identification tag or custody seal
Specifications
Capacity16 oz., 500mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorWhite, Translucent
DescriptionSquare Bottle HDPE 500mL
Diameter (English) Inner Neck1.7 in.
Diameter (Metric) Outer Neck45 mm (Inner Neck)
Diameter (Metric) Inner Neck45 mm
IncludesInternal Plug
MaterialHDPE
Length (Metric)70mm
Max. Temperature (Metric)110°C
Max. Temperature (English)230°F
Neck ID (Metric)45mm
No. per Case36
No. per Pack6
QuantityCase, Pack
SeptaYes
SterilityNon-sterile
TransparencyTranslucent
Width (Metric)80mm
Closure ColorTranslucent
Graduations50mL
Closure IncludedYes
GraduatedYes
MouthWide
ShapeSquare
Height (Metric)138 mm
Product TypeSquare Bottle
Width (English)3.15 in.
Length (English)2.76 in.
Height (English)5.38 in.
Closure MaterialHDPE
Closure TypeScrew Cap
Through the Closure DispensingNo