Pycnometer
Pycnometer
Actual product may vary
Pycnometer
Pycnometer
Pycnometer
Pycnometer
For specific gravity determinations of semifluids
Have Questions?
Catalog number FIS03-247Q
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For specific gravity determinations of semifluids
  • Three-part aluminum alloy construction
  • Withstands pressure needed to expel air from viscous material and fill pycnometer completely
  • Three-part construction allows close manufacturing tolerances, prevents material under test from being forced into threads
  • Parts are cut from solid bar stock to avoid pits or cavities
Specifications
Diameter (Metric)50 mm
Capacity (Metric)11.5 mL
Height (Metric)25 mm
Diameter (English)2 in.
MaterialAluminum Alloy
Height (English)1 in.
For Use With (Application)Measure the specific gravity of semifluids
Unit SizeEA