Polypropylene Graduated Cylinders
Actual product may vary
Polypropylene Graduated Cylinders
Calibrated To Contain/To Deliver at 20°C.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FIS03-007-42250 mL2 mL
Catalog number FIS03-007-42
Price (SGD)
13.00
Each
Capacity (Metric):
250 mL
Graduations:
2 mL
Price (SGD)
13.00
Each
Calibrated To Contain/To Deliver at 20°C
  • Translucent cylinders
  • Molded-in large, easy-to-read graduations
  • Large rounded base ribbed for reinforcement and stability
  • Generous pour spout; 10 mL size has a conical top for easier filling and pouring
  • Non-wetting
  • Chemical resistant
  • Contact-clear
Specifications
Capacity (Metric)250 mL
MaterialPolypropylene
Certifications/ComplianceASTM
CalibrationTo Contain, To Deliver
Graduations2 mL
AutoclavableAutoclavable
Subdivision2mL
ShapeRound
Base TypeCircular
Product TypeGraduated Cylinder
Unit SizeEach