Polypropylene Graduated Cylinders
Actual product may vary
Fisherbrand
Polypropylene Graduated Cylinders
Calibrated To Contain/To Deliver at 20°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS03-007-3810 mL0.2 mL
FIS03-007-4050 mL1 mL
FKS1196788450 mL1 mL
FKS1194788410 mL0.2 mL
FIS03-007-3925 mL0.5 mL
FKS1195788425 mL0.5 mL
FIS03-007-41100 mL1 mL
FIS03-007-42250 mL2 mL
FKS11907884250 mL2 mL
FIS03-007-43500 mL5 mL
FKS11967874500 mL5 mL
FIS03-007-441000 mL10 mL
FKS119378741000 mL10 mL
FIS03-007-452000 mL20 mL
FKS119278742000 mL20 mL
Catalog number FIS03-007-38
Price (SGD)
6.00
Each
Capacity (Metric):
10 mL
Graduations:
0.2 mL
Price (SGD)
6.00
Each
Calibrated To Contain/To Deliver at 20°C
  • Translucent cylinders
  • Molded-in large, easy-to-read graduations
  • Large rounded base ribbed for reinforcement and stability
  • Generous pour spout; 10 mL size has a conical top for easier filling and pouring
  • Non-wetting
  • Chemical resistant
  • Contact-clear
Specifications
Capacity (Metric)10 mL
MaterialPolypropylene
Certifications/ComplianceASTM
CalibrationTo Contain, To Deliver
Graduations0.2 mL
AutoclavableAutoclavable
Subdivision0.2mL
ShapeRound
Base TypeCircular
TypeGraduated Cylinder
Unit SizeEach