Entry Level Research Grade Inverted Microscope
Actual product may vary
Entry Level Research Grade Inverted Microscope
Entry Level Research Grade Inverted Microscope
Entry Level Research Grade Inverted Microscope
Entry Level Research Grade Inverted Microscope
Entry Level Research Grade Inverted Microscope
Entry Level Research Grade Inverted Microscope
Fisherbrand
Entry Level Research Grade Inverted Microscope
Entry Level Research Grade Inverted Microscope is a simple, easy-to-use entry level inverted microscope, ideal for quickly checking cell health
Have Questions?
Catalog NumberObjective Types
FIS03-000-013Infinity Corrected Plan Achromatic, Plan Phase
Catalog number FIS03-000-013
Price (SGD)
9,005.00
Each
Objective Types:
Infinity Corrected Plan Achromatic, Plan Phase
Price (SGD)
9,005.00
Each
OPTICAL SYSTEM
  • Infinity optical system
  • Backward facing 5 position turret nosepiece w/ click stops
  • LWD 4x PL BF, & 10X, 20X LWD PL PH objectives
  • 10X WF w/ 22mm FOV eyepiece
  • Corrected for glasses
  • Full Koehler illumination
VIEWING HEAD
  • Siedentopf Trinocular head with 30° incline
  • 82/20 split between camera and eyepieces
  • Dual eyepiece diopter adjustment -8° to +5°
  • Interpupillary adjustment distance of 48mm to 76mm
  • Ability to attach a camera
FOCUSING
  • Coaxial coarse and fine focusing adjustment
  • Tension adjustments
  • Fine focus drive movement of 0.001mm divisions
  • Moving range: 24 mm
  • Rack stop adjustment for protecting objective and specimen slides from accidental breakage
CONDENSER
  • 72 mm WD Abbe Condenser w/ Phase slider
  • Removable Condenser allows for up to 195 mm WD to accommodate large vessels
  • NA 0.30 with iris diaphragm
STAGE
  • Size: 240 mm (x) x 210 mm (y)
FRAME
  • Rugged, durable metal frame construction
  • Rocker on/off switch
ILLUMINATION
  • Eco-friendly and energy efficient 5W LED illumination
  • Dimmer control for light intensity
Specifications
Condenser TypePhase contrast condenser
Design TypeInfinite Optical System
Eyepiece10X/22 mm
Head StyleSiedentopf Trinocular, 30° Inclined
Lighting TypeLED, Adjustable
Magnification Power4X, 10X and 20X
ModelInfinity Optical System
Objective TypesInfinity Corrected Plan Achromatic, Plan Phase
Product TypeMicroscope
IncludesEyepieces, Stand, Dust Cover, Objectives, Power Cord
MaterialBrass, Glass, Aluminum, Plastic
Optical DesignInfinity Optical System
Warranty5 years optical and mechanical, 1 year electrical from date of purchase
Electrical Requirements110/220 V
Voltage110/220 V